missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Stainless Steel Receiver For Test Sieves

For the collection of materials processed through Fisherbrand™ Stainless Steel Test Sieves.

Brand:  Fisherbrand™ 01/200SBWREC

 View more versions of this product

Product Code. 11517033

  • 38.66 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Available for 100 and 200mm diameter sizes
  • Conforms to BS 410: Part 1: 2000, ISO 3310: Part 1: 2000
Specifications

Specifications

1 Each
Stainless Steel
200 mm
Sieve Receiver
Collection of sieved materials
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.