missing translation for 'onlineSavingsMsg'
Learn More

De Buyer™ Stainless Steel Serving Dishes

Brand:  De Buyer™ 3280.35

 View more versions of this product

Product Code. 11990615

  • 91.00 € / Each
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Dish
45 mm
Each
3.6 L
Stainless Steel
350 mm
Rectangular
270 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers