missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh

Made from stainless steel and available in a wide variesty of sizes. Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh has an 100 mm diameter.

Brand:  Fisherbrand™ 01/100SBW2.80mm

 View more versions of this product

Product Code. 11552173

  • 65.35 € / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

100 mm diameter test sieves with stainless steel frame and woven wire mesh. Supplied with Certificate of Conformity and record card. Additional mesh sizes and diameter sieves available - please contact Customer Service for details.
Specifications

Specifications

1 Ea.
Sieve
Stainless Steel
100 mm
Woven Wire
2.8 mm
Certificate of Conformity, BS 410: Part 1: 2000, ISO 3310: Part 1: 2000
Certificate of conformity and record card
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.