Learn More
RETSCH Stainless-Steel Test Sieve, 200 Dia. x 25 mmH, Pore sizes in micrometers, ISO 3310/1
Use for separation, fractioning, or particle size determination. These stainless-steel test sieves have an internal height of 25 mm and diameter of 200 mm. Available in a wide range of micrometer pore sizes.
114.00 € - 831.00 €
Specifications
Quantity | 1 Each |
---|---|
Type | Test Sieve |
For Use With (Equipment) | Shaker AS 200 |
Height (Metric) | 25mm |
For Use With (Application) | For separation, fractioning, particle size determination |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
10564872
|
RETSCH
60.122.000106 |
106 μm |
114.00 €
Each |
Estimated Shipment: 07-03-2025
Log in to see stock. |
|||||
10761233
|
RETSCH
60.122.000112 |
112 μm |
114.00 €
Each |
Estimated Shipment: 07-03-2025
Log in to see stock. |
|||||
10186211
|
RETSCH
60.122.000140 |
140 μm |
118.00 €
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
|||||
10739512
|
RETSCH
60.122.000160 |
160 μm |
114.00 €
Each |
Estimated Shipment: 07-03-2025
Log in to see stock. |
|||||
10596212
|
RETSCH
60.122.000200 |
200 μm |
119.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
10146211
|
RETSCH
60.122.000020 |
20 μm |
831.00 €
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
|||||
10196451
|
RETSCH
60.122.000212 |
212 μm |
118.00 €
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
|||||
10014921
|
RETSCH
60.122.000224 |
224 μm |
118.00 €
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
|||||
10330182
|
RETSCH
60.122.000025 |
25 μm |
437.00 €
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
|||||
10699911
|
RETSCH
60.122.000280 |
280 μm |
114.00 €
Each |
Estimated Shipment: 07-03-2025
Log in to see stock. |
|||||
10298921
|
RETSCH
60.122.000300 |
300 μm |
118.00 €
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
|||||
10630102
|
RETSCH
60.122.000315 |
315 μm |
114.00 €
Each |
Estimated Shipment: 07-03-2025
Log in to see stock. |
|||||
10238491
|
RETSCH
60.122.000032 |
32 μm |
258.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
10496231
|
RETSCH
60.122.000355 |
355 μm |
114.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
10574672
|
RETSCH
60.122.000036 |
36 μm |
245.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
10330862
|
RETSCH
60.122.000400 |
400 μm |
119.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
10701233
|
RETSCH
60.122.000038 |
38 μm |
245.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
10460022
|
RETSCH
60.122.000040 |
40 μm |
172.00 €
Each |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||||
10125981
|
RETSCH
60.122.000425 |
425 μm |
118.00 €
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
|||||
10135981
|
RETSCH
60.122.000450 |
450 μm |
118.00 €
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
|||||
10349511
|
RETSCH
60.122.000045 |
45 μm |
177.00 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||||
10106311
|
RETSCH
60.122.000050 |
50 μm |
154.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
10288921
|
RETSCH
60.122.000053 |
53 μm |
154.00 €
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
|||||
10258781
|
RETSCH
60.122.000560 |
560 μm |
121.00 €
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
|||||
10034801
|
RETSCH
60.122.000056 |
56 μm |
137.00 €
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
|||||
10690102
|
RETSCH
60.122.000600 |
600 μm |
118.00 €
Each |
Estimated Shipment: 07-03-2025
Log in to see stock. |
|||||
15630369
|
RETSCH
60.122.000630 |
630 μm |
118.00 €
Each |
Estimated Shipment: 07-03-2025
Log in to see stock. |
|||||
10447581
|
RETSCH
60.122.000710 |
710 μm |
118.00 €
Each |
Estimated Shipment: 07-03-2025
Log in to see stock. |
|||||
10054801
|
RETSCH
60.122.000071 |
71 μm |
118.00 €
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
|||||
10711623
|
RETSCH
60.122.000075 |
75 μm |
114.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
10690082
|
RETSCH
60.122.000080 |
80 μm |
114.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
10259021
|
RETSCH
60.122.000850 |
850 μm |
121.00 €
Each |
Estimated Shipment: 28-03-2025
Log in to see stock. |
|||||
10065061
|
RETSCH
60.122.000900 |
900 μm |
123.00 €
Each |
Estimated Shipment: 07-03-2025
Log in to see stock. |
|||||
10600092
|
RETSCH
60.122.000090 |
90 μm |
114.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
10461973
|
RETSCH
60.122.000180 |
180 μm |
114.00 €
Each |
Estimated Shipment: 07-03-2025
Log in to see stock. |
|||||
Description
- Stainless-steel frame with wire gauze
- Diameter: 200 mm
- Height: 25 mm
- ISO 3310/1
Specifications
1 Each | |
Shaker AS 200 | |
For separation, fractioning, particle size determination | |
Stainless Steel |
Test Sieve | |
25mm | |
200 mm | |
Stainless Steel |