Learn More
Fisherbrand™ Stainless-Steel Test Sieves, 200 Dia. x 25 mmH, ISO 3310/1
Use for separation, fractioning, or particle size determination. These stainless-steel test sieves have an internal height of 25 mm and diameter of 200 mm. Available in a wide range of pore sizes between 1mm and 10 mm.
102.00 € - 173.00 €
Specifications
Quantity | 1 Each |
---|---|
Type | Test Sieve |
For Use With (Equipment) | Shaker AS 200 |
Height (Metric) | 25mm |
For Use With (Application) | For separation, fractioning, particle size determination |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
10208551
|
Fisherbrand™
60.122.001180 |
1.18 mm |
126.00 €
Each |
Estimated Shipment: 19-03-2025
Log in to see stock. |
|||||
10789702
|
Fisherbrand™
200SBIW1.25H |
1.25 mm | N/A | ||||||
10466811
|
Fisherbrand™
60.122.001120 |
1.12 mm |
122.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
10789892
|
Fisherbrand™
60.122.001400 |
1.4 mm |
122.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
10340482
|
Fisherbrand™
60.122.001600 |
1.6 mm |
126.00 €
Each |
Estimated Shipment: 19-03-2025
Log in to see stock. |
|||||
10585832
|
Fisherbrand™
200SBIW1.70H |
1.7 mm | N/A | ||||||
10044891
|
Fisherbrand™
60.122.010000 |
10 mm |
173.00 €
Each |
Estimated Shipment: 28-02-2025
Log in to see stock. |
|||||
10595832
|
Fisherbrand™
60.122.001800 |
1.8 mm |
122.00 €
Each |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||||
10065071
|
Fisherbrand™
200SBIW1.00H |
1 mm | N/A | ||||||
10298931
|
Fisherbrand™
60.122.002240 |
2.24 mm |
126.00 €
Each |
Estimated Shipment: 19-03-2025
Log in to see stock. |
|||||
10792003
|
Fisherbrand™
60.122.002360 |
2.36 mm |
122.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
10534112
|
Fisherbrand™
60.122.002500 |
2.5 mm |
122.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
10126981
|
Fisherbrand™
60.122.002800 |
2.8 mm |
154.00 €
Each |
Estimated Shipment: 19-03-2025
Log in to see stock. |
|||||
10196551
|
Fisherbrand™
200SBIW2.00H |
2 mm | N/A | ||||||
10780663
|
Fisherbrand™
60.122.003150 |
3.15 mm |
151.00 €
Each |
In Stock
Log in to see stock. |
|||||
10005081
|
Fisherbrand™
60.122.003350 |
3.35 mm |
154.00 €
Each |
Estimated Shipment: 19-03-2025
Log in to see stock. |
|||||
10610462
|
Fisherbrand™
60.122.004500 |
4.5 mm |
151.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
10258641
|
Fisherbrand™
60.122.004750 |
4.75 mm |
154.00 €
Each |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
10573532
|
Fisherbrand™
60.122.003550 |
3.55 mm |
151.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
10248641
|
Fisherbrand™
60.122.004000 |
4 mm |
154.00 €
Each |
Estimated Shipment: 28-02-2025
Log in to see stock. |
|||||
10379651
|
Fisherbrand™
60.122.005600 |
5.6 mm |
154.00 €
Each |
Estimated Shipment: 28-02-2025
Log in to see stock. |
|||||
10389651
|
Fisherbrand™
60.122.006300 |
6.3 mm |
154.00 €
Each |
Estimated Shipment: 19-03-2025
Log in to see stock. |
|||||
10649691
|
Fisherbrand™
60.122.005000 |
5 mm |
167.00 €
Each |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
10729122
|
Fisherbrand™
60.122.006700 |
6.7 mm |
151.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
10319801
|
Fisherbrand™
60.122.007100 |
7.1 mm |
154.00 €
Each |
Estimated Shipment: 19-03-2025
Log in to see stock. |
|||||
10629881
|
Fisherbrand™
200SBIW8.00H |
8 mm | N/A | ||||||
10329801
|
Fisherbrand™
60.122.009000 |
9 mm |
154.00 €
Each |
Estimated Shipment: 19-03-2025
Log in to see stock. |
|||||
10176591
|
Fisherbrand™
60.122.009500 |
9.5 mm |
154.00 €
Each |
Estimated Shipment: 28-02-2025
Log in to see stock. |
|||||
Description
- Stainless-steel frame with wire gauze
- Diameter: 200 mm
- Height: 25 mm
- ISO 3310/1
Specifications
1 Each | |
Shaker AS 200 | |
For separation, fractioning, particle size determination | |
Stainless Steel |
Test Sieve | |
25mm | |
200 mm | |
Stainless Steel |