missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50 mmH, with Perforated Plates with Round Holes and Pore Size Greater than 10 mm
Brand: RETSCH 60.198.022400
This item is not returnable.
View return policy
Specifications
1 Each | |
Test Sieve | |
Certificate indicating Compliance | |
For separation, fractioning, particle size determination | |
Stainless Steel |
22.4 mm | |
ISO 3310/2 | |
50mm | |
200 mm | |
Stainless Steel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction