missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH Stainless-Steel Test Sieves, 200 mm Dia., with Calibration Certificate
Brand: RETSCH 60.331.000500
This item is not returnable.
View return policy
Specifications
1 Each | |
Test Sieve | |
50mm | |
200 mm | |
Stainless Steel |
500 μm | |
ISO 3310/1 and ASTM E 11 | |
Separation, fractioning, particle size determination | |
Stainless Steel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction