Learn More
RETSCH Stainless-Steel Test Sieves, 450 mm Dia., in Pore Sizes Greater than 10 mm with ISO 3310/1
687.00 € - 1120.00 €
Specifications
Quantity | 1 Each |
---|---|
Type | Test Sieve |
Certifications/Compliance | ISO 3310/1 |
For Use With (Equipment) | Vibratory Shaker AS 450 |
Includes | Certificate indicating Compliance |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
11904632
|
RETSCH
60.168.100000 |
100 mm |
708.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11914632
|
RETSCH
60.168.106000 |
106 mm |
708.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11954602
|
RETSCH
60.168.011200 |
11.2 mm |
687.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11924632
|
RETSCH
60.168.112000 |
112 mm |
708.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11964602
|
RETSCH
60.168.012500 |
12.5 mm |
687.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11934632
|
RETSCH
60.168.125000 |
125 mm |
708.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11974602
|
RETSCH
60.168.013200 |
13.2 mm |
687.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11984602
|
RETSCH
60.168.014000 |
14 mm |
687.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11994602
|
RETSCH
60.168.016000 |
16 mm |
687.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11904612
|
RETSCH
60.168.018000 |
18 mm |
687.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11914612
|
RETSCH
60.168.019000 |
19 mm |
687.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11924612
|
RETSCH
60.168.020000 |
20 mm |
687.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11934612
|
RETSCH
60.168.022400 |
22.4 mm |
687.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11944612
|
RETSCH
60.168.025000 |
25 mm |
1120.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11954612
|
RETSCH
60.168.026500 |
26.5 mm |
687.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11964612
|
RETSCH
60.168.028000 |
28 mm |
708.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11974612
|
RETSCH
60.168.031500 |
31.5 mm |
708.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11984612
|
RETSCH
60.168.035500 |
35.5 mm |
708.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11994612
|
RETSCH
60.168.037500 |
37.5 mm |
708.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11904622
|
RETSCH
60.168.040000 |
40 mm |
708.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11914622
|
RETSCH
60.168.045000 |
45 mm |
708.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11934622
|
RETSCH
60.168.053000 |
53 mm |
708.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11944622
|
RETSCH
60.168.056000 |
56 mm |
708.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11924622
|
RETSCH
60.168.050000 |
50 mm |
708.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11954622
|
RETSCH
60.168.063000 |
63 mm |
708.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11964622
|
RETSCH
60.168.071000 |
71 mm |
708.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11974622
|
RETSCH
60.168.075000 |
75 mm |
708.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
11984622
|
RETSCH
60.168.080000 |
80 mm |
708.00 €
Each |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||||
11994622
|
RETSCH
60.168.090000 |
90 mm |
708.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
Specifications
1 Each | |
ISO 3310/1 | |
Certificate indicating Compliance | |
For separation, fractioning, particle size determination | |
Stainless Steel |
Test Sieve | |
Vibratory Shaker AS 450 | |
100mm | |
450 mm | |
Stainless Steel |