missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Stainless Steel Collecting Pan for Test Sieves

Collects resulting samples from test sieves. These Stainless Steel pans are compatible with multiple test sieves and sieve shakers, based upon the required diameter.

Brand:  RETSCH 60.010.000305

132.05 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10790843

  • 143.00 € / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stainless Steel
  • Compatible with multiple test sieves, based upon diameter
Specifications

Specifications

Collecting Pan
Test Sieve
305 mm
Stainless Steel
40 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers