missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Stainless Steel Test Tube Rack

Fits a variety of test tube sizes from 10 to 30mm diameter

Brand:  Grant Instruments™ SR-30

 View more versions of this product

Product Code. 15365842

  • 182.00 € / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Fits a variety of test tube sizes from 10 to 30mm diameter

  • For use with LSB Aqua Pro
  • Available in 8 sizes to hold up to 48 SR test tubes
Specifications

Specifications

10 mL
30mm dia. Tubes
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers