Learn More
Grant Instruments™ Stainless Steel Test Tube Rack
Fits a variety of test tube sizes from 10 to 30mm diameter
169.60 € - 182.00 €
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15385842
|
Grant Instruments™
SR-SE |
0.5mm dia. Tubes | N/A | ||||||
15365832
|
Grant Instruments™
SR-10 |
10mm dia. Tubes |
182.00 €
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||||
15385832
|
Grant Instruments™
SR-13 |
13mm dia. Tubes |
182.00 €
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
|||||
15305842
|
Grant Instruments™
SR-16 |
16mm dia. Tubes |
182.00 €
Each |
Estimated Shipment: 07-03-2025
Log in to see stock. |
|||||
15325842
|
Grant Instruments™
SR-19 |
19mm dia. Tubes |
182.00 €
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||||
15395842
|
Grant Instruments™
SR-LE |
1.5mm dia. Tubes |
182.00 €
Each |
Estimated Shipment: 07-03-2025
Log in to see stock. |
|||||
15345842
|
Grant Instruments™
SR-25 |
25mm dia. Tubes |
182.00 €
Each |
Estimated Shipment: 07-03-2025
Log in to see stock. |
|||||
15365842
|
Grant Instruments™
SR-30 |
30mm dia. Tubes |
182.00 €
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
|||||
Description
Fits a variety of test tube sizes from 10 to 30mm diameter
- For use with LSB Aqua Pro
- Available in 8 sizes to hold up to 48 SR test tubes
Specifications
Stainless Steel |