Learn More
Grant Instruments™ Stainless Steel Test Tube Rack
Fits a variety of test tube sizes from 10 to 30mm diameter
Brand:  Grant Instruments™ SR-SE
 View more versions of this product
Product Code. 15385842
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Fits a variety of test tube sizes from 10 to 30mm diameter
  • For use with LSB Aqua Pro
  • Available in 8 sizes to hold up to 48 SR test tubes
Specifications
Specifications
119 mL
0.5mm dia. Tubes
Stainless Steel