Learn More
Bochem™ Stainless Steel Wire Basket, Square, 2 Cross-ties
Place laboratory items in the Bochem™ Stainless Steel Wire Basket, Square, 2 Cross-ties. .
Brand:  Bochem™ 10076
 View more versions of this product
Product Code. 10234271
  • 263.00 € / Each
Estimated Shipment: 25-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Mesh width 8x8x1mm,
  • Square
  • Stainless Steel
  • 2 cross-ties
Specifications
Specifications
Wire Basket
600 mm
600 L x 400 Wmm
600 x 400 x 300 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bochem™ Stainless Steel Wire Basket, Square, 2 Cross-ties > Dimensions (L x W x H): 600 x 400 x 300mm