missing translation for 'onlineSavingsMsg'
Learn More

X10 Standard Solution, 10mL

Provides easy calibration of Osmometers. Loeser™ Standard Solution Standard ships 10 glass ampoules of 1mL content.

12.25 € - 26.25 €

Specifications

Packaging Glass ampoules
Quantity 10 x 1 mL
pH 7
View More Specs

Products 2
Product Code Brand Concentration Melting Point Boiling Point Price Quantity & Availability  
Product Code Brand Concentration Melting Point Boiling Point Price Quantity & Availability  
11970088
View Documents
Loeser
0.01.0001
300mosm/kg H2O -0.55°C 100.4°C
26.25 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
11920557
View Documents
Loeser
0.01.0002
900mosm/kg H2O -1672°C 101°C
12.25 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Standard Solution for Osmometer
  • Pack of 10 glass ampoules of 1mL content.
  • For easy calibration of Osmometers
Specifications

Specifications

Glass ampoules
10 x 1 mL
7
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.