Learn More
Cole-Parmer™ Standard Digital Gloss Meter
Use this gloss meter to evaluate the gloss of any coated surface, check paints or inks, inspect print materials, and more.
Brand:  Cole-Parmer™ 59600-05
Product Code. 13476503
Description
Description
  • Using an infrared source that emits a beam of light at an angle to the surface, a sensor measures the reflected light.
  • Features a 3-digit LCD and is switch selectable for a standard 60° measurement angle or a 20° angle for measuring high-gloss surfaces.
  • The sensor is separate from the meter for measuring hard-to-reach surfaces.
  • Includes 1-ft (30.5 cm) coiled connection cable and four 1.5 V AAA batteries.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Standard Digital Gloss Meter > Voltage: 1.5V