Learn More
Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco™ Injectors
Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco Injectors offer easy, accurate and reproducible manual injections.
Brand:  Thermo Scientific™ 365IL271
 View more versions of this product
Product Code. 10120553
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco Injectors offer easy, accurate and reproducible manual injections.
  • Square tip to prevent damage to the injector
  • Wide range of volumes
  • Precision made from borosilicate glass and stainless steel
  • Robust design and easy-to-read markings
TRUSTED_SUSTAINABILITY
Specifications
Specifications
250uL
Reusable
Injector
HPLC Autosampler Syringe
250 μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.