Learn More
Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco™ Injectors
Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco Injectors offer easy, accurate and reproducible manual injections.
49.03 € - 100.93 €
Specifications
Product Type | HPLC Autosampler Syringe |
---|---|
Disposable | Reusable |
For Use With (Equipment) | Injector |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10324133
|
Thermo Scientific™
365CL221 |
5 μL | N/A | ||||||
10110743
|
Thermo Scientific™
365DL231 |
10 μL | N/A | ||||||
10715564
|
Thermo Scientific™
365FL241 |
25 μL | N/A | ||||||
10171073
|
Thermo Scientific™
365GL251 |
50 μL |
65.55 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10160503
|
Thermo Scientific™
365HL261 |
100 μL | N/A | ||||||
10120553
|
Thermo Scientific™
365IL271 |
250 μL | N/A | ||||||
10631714
|
Thermo Scientific™
365JL281 |
500 μL |
100.93 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco Injectors offer easy, accurate and reproducible manual injections.
- Square tip to prevent damage to the injector
- Wide range of volumes
- Precision made from borosilicate glass and stainless steel
- Robust design and easy-to-read markings
Specifications
HPLC Autosampler Syringe | |
Reusable | |
Injector |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.