missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Standard Fluid Transfer Assembly Y Set

Simplify process connectivity and enable greater efficiency for most upstream and downstream process operations.

286.62 € - 960.00 €

Specifications

Content And Storage Approved for storage at ambient conditions (15°C to 30°C).
View More Specs

Products 5
Product Code Brand Tubing Style Price Quantity & Availability  
Product Code Brand Tubing Style Price Quantity & Availability  
17840627
View Documents
Thermo Scientific™
SH31092.01
3 x 3.2mm (1/8 in) ID x 1.6mm (1/16 in) wall, C-Flex 374, 2 x 0.51m (20 in) to 0.51m (20 in)
345.00 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
17850627
View Documents
Thermo Scientific™
SH31093.01
3 x 6.35mm (1/4 in) ID x 1.6mm (1/16 in) wall, C-Flex 374, 2 x 0.51m (20 in) to 0.51m (20 in)
861.00 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
17800807
View Documents
Thermo Scientific™
SH31094.01
3 x 6.35mm (1/4 in) ID x 2.4mm (3/32 in) wall, C-Flex 374, 2 x 0.51m (20 in) to 0.51m (20 in) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17890677
View Documents
Thermo Scientific™
SH31095.01
3 x 9.52mm (3/8 in) ID x 3.2mm (1/8 in) wall, C-Flex 374, 2 x 0.51m (20 in) to 0.51m (20 in)
960.00 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
17880627
View Documents
Thermo Scientific™
SH31096.01
3 x 12.7mm (1/2 in) ID x 3.2mm (1/8 in) wall, C-Flex 374, 2 x 0.51m (20 in) to 0.51m (20 in)
486.00 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.

  •  One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications

Specifications

Approved for storage at ambient conditions (15°C to 30°C).
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Standard Fluid Transfer Assembly Y Set

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.