Learn More
Sampling Systems™ Standard Steel Sample Splitter Riffle Boxes
These easy-to-use Sample Splitters in standard steel are for dividing and reducing bulk samples.
Marca:  Sampling Systems™ 8600M-15
 vedi altre versioni di questo prodotto
Codice prodotto. 15889521
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi »
Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Simply drop the sample into the top and the sample is split in two
  • Split sample drops into the riffle boxes underneath
  • It is recommended that width of slots are 2.5 to 3 times bigger than the diameter of the largest particle being split
  • Available in a wide range of sizes
Specifica
Specifica
Riffle Box
5.13 L
Dividing and Reducing Bulk Samples
15 mm
Steel
12
232 x 91 x 122 mm
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documenti
Documenti
Certificati
Offerte speciali
Offerte speciali