missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cut Out Rubber Covered Rings

With cut out section to allow easy removal of equipment.

Brand:  Fisherbrand™ 2015/6

 View more versions of this product

Product Code. 11735626

  • 25.25 € / Each
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • With rubber covered rings
  • Stem length to the center of bosshead is approximately 60mm
  • Constructed in mild steel with welded joints
  • Zinc-plated

Specifications

Specifications

Mild Steel
Stativring
Zinc Plated, Rubber Coated
75 mm
Cut out section
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.