missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cut Out Rubber Covered Rings

With cut out section to allow easy removal of equipment.

Brand:  Fisherbrand™ 2015/14

 View more versions of this product

Product Code. 11755626

  • 23.60 € / Each
Estimated Shipment: 08-10-2024
to see stock.
Explore more special offers
Add to basket

Description

Description

  • With rubber covered rings
  • Stem length to the center of bosshead is approximately 60mm
  • Constructed in mild steel with welded joints
  • Zinc-plated

Specifications

Specifications

Mild Steel
Stativring
Zinc Plated, Rubber Coated
125 mm
Cut out section
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers