Learn More
Fisherbrand™ Stainless Steel Straight Tongs
Get maximum results using laboratory tongs in high temperature chemical reactions. Made of durable stainless steel, these Clifton™ Stainless Steel Straight Tongs ensure precise control of grasping, lifting, turning, and transferring heat-resistant and other materials in a laboratory environment.
8.80 € - 11.30 €
Specifications
Material Steel
Type Crucible Tongs
Quantity 1 Ea.
View More Specs
Products 2
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
11593492
View Documents
Fisherbrand™
3030
150 mm
8.80 €
Each
In Stock
to see stock.
Edge
Add to basket
11503502
View Documents
Fisherbrand™
3031
200 mm
11.30 €
Each
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Laboratory straight tongs
  • Stainless steel 304 spec 18/8 non-magnetic, bright finish
  • Designed for corrosion resistance and durability ideal in laboratory routines
  • Laboratory metalware consumables
  • Finest in corrosion-resistant 304 stainless steel
  • Provides corrosion resistance that prevents product contamination
Specifications
Specifications
Steel
Crucible Tongs
1 Ea.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Stainless Steel Straight Tongs