Learn More
Fisherbrand™ Stainless Steel Straight Tongs
Get maximum results using laboratory tongs in high temperature chemical reactions. Made of durable stainless steel, these Clifton™ Stainless Steel Straight Tongs ensure precise control of grasping, lifting, turning, and transferring heat-resistant and other materials in a laboratory environment.
Brand:  Fisherbrand™ 3030
 View more versions of this product
Product Code. 11593492
  • 8.80 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Laboratory straight tongs
  • Stainless steel 304 spec 18/8 non-magnetic, bright finish
  • Designed for corrosion resistance and durability ideal in laboratory routines
  • Laboratory metalware consumables
  • Finest in corrosion-resistant 304 stainless steel
  • Provides corrosion resistance that prevents product contamination
Specifications
Specifications
1 Ea.
150 mm
Steel
Crucible Tongs
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Stainless Steel Straight Tongs > Length: 150mm