missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Steel Disposal Bin

The Steel Disposal Bin is ideal for the safe collection of solvent-soaked cleaning rags and other waste.

Brand:  asecos™ 23158

 View more versions of this product

Product Code. 17856426

  • 274.00 € / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Sheet Steel
  • Disposal bin ideal for safe disgard of solvant-soaked rags.
  • Self-closing lid with damping, large, ergonomically shaped handle on the rear of the container for easy emptying of the container
  • Opening by large pedal with anti-slip surface
  • Available in various capacities.
Specifications

Specifications

65 L
535.00 mm
Red
21.83 lb.
545.00 mm
22.83 in.
Liquid Disposal Can
21.06 in.
17.2 gal.
580.00 mm
9.9 kg
21.46 in.
Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.