missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Steel Disposal Bin
The Steel Disposal Bin is ideal for the safe collection of solvent-soaked cleaning rags and other waste.
167.00 € - 274.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
17866486
|
asecos™
39379 |
20 L |
167.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
17876546
|
asecos™
23156 |
35 L |
186.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
17836496
|
asecos™
23157 |
50 L |
237.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
17856426
|
asecos™
23158 |
65 L |
274.00 €
Each |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
Description
- Sheet Steel
- Disposal bin ideal for safe disgard of solvant-soaked rags.
- Self-closing lid with damping, large, ergonomically shaped handle on the rear of the container for easy emptying of the container
- Opening by large pedal with anti-slip surface
- Available in various capacities.
Specifications
Red | |
Steel | |
Liquid Disposal Can |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title