Learn More
R&L Enterprises™ Plated Steel Spring
An excellent option for hand use or bosshead mounting. R&L Enterprises™ Plated Steel Springs provide a gentle yet firm grip on laboratory tubing.
Brand:  R&L Enterprises™ Z5904
Product Code. 13078423
  • 19.65 € / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • With alloy rod
  • For tubes up to 30mm in diameter
  • Supplied in packs of 10
Specifications
Specifications
10 Pk.
Steel
Test Tube Holder Spring
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&L Enterprises™ Plated Steel Spring > Material: Steel