missing translation for 'onlineSavingsMsg'
Learn More

R&L Enterprises™ Steel Wire Gauze

Provides excellent support to beakers and other glassware during heating on ring stands and tripods. R&L Enterprises™ Steel Wire Gauze is a flat square manufactured from steel mesh.

Brand:  R&L Enterprises™ 9076

 View more versions of this product

Product Code. 12968950

  • 30.52 € / Pack of 10
Estimated Shipment: 26-06-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Non-fraying edges
  • Durable and long-lasting
  • Excellent heat distribution
Specifications

Specifications

Ceramic Center
Non-Fraying
Square
Steel
150 mm
150 x 150 mm
10 Pack
150 mm
Wire Gauze
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
R&L Enterprises™ Steel Wire Gauze > Dimensions: 150 x 150mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.