Learn More
Zeiss™ Stemi 305 Stereo Microscope
Compact Greenough stereo microscope with 5:1 zoom for biological education, labs and production environments.
Brand: Zeiss™ 435063-9030-100
1812.60 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- This compact Greenough stereo microscope allows you to observe your samples as they are: three-dimensional and crisp in contrast and no preparation required.
- The Stemi 305 is an easy-to-use microscope, requires minimal setup, everything is integrated including long-living LED illumination for both reflected and transmitted light.
- Spot Illuminator K LED - Single spot for brightfield and darkfield epi-illumination, Thread M24x0.5 for polarizer, Height adjustable, inclinable, zoomable.
Specifications
Ring Illuminator K LED | |
Stereo Microscope |