Learn More
Contec™ Sterile Denatured Ethanol, 70%

Blend of 70% v/v denatured ethanol with 30% purified water. Contec™ Sterile Low Endotoxin Alcohol, 70% is 0.2 micron filtered.
Brand: Contec™ SBT0570DEW
194.46 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.
Description
- Guaranteed endotoxin levels of <0.25EU/mL make it ideal for use in product contact areas
- GMP manufactured under Grade A laminar flow air flow in a Grade C cleanroom ensures the alcohol, container and packaging are free from contamination and particulates
- Trigger spray and bag in bottle protected system prevents bottle contents from becoming contaminated during use
- Bottles can be completely emptied eliminating wastage so no need to discard unused product during shelf life
- Trigger spray can be set to jet or spray - large droplet size reduces the risk of inhalation and provides good surface coverage
- Double/triple bagged packed in linear tear packaging so each bag is easy to open even when wearing gloves
- Sterile via gamma irradiation so suitable for use in Grade A and B cleanrooms

Specifications
Denatured Ethanol with WFI | |
Not Applicable | |
70% Denatured Ethanol in 30% WFI | |
8 x 0.5 L | |
0.5 L |
Liquid Spray | |
Sterile | |
Trigger Spray Bottle | |
0.1 gal. |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- CONTEC STERILE DENATURED ETHANOL WITH WFI
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use dry chemical/coarbindioxide to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P411-Store at temperatures not exceeding 40°C/°F.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol, propan-2-ol
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.