missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Sterile Graduated Transfer Pipets
Description
- Transparent in color
- Contains graduations
- Sterile
Specifications
LDPE | |
30 | |
500/Pk. | |
25 inner packs of 20 | |
150 mm |
Sterile | |
Yes | |
33 μL | |
1 mL | |
Transfer Pipet |
Fisherbrand Focus on glassware and plasticware
File size: 66.4 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Sterile Graduated Transfer Pipets > Capacity: 1mL; Drops/mL: 30; 20 Case
Spot an opportunity for improvement?Share a Content Correction