Learn More
Technical Service Consultants™ Sterile Inoculation Loop
Use the Technical Service Consultants™ Sterile Inoculation Loop for general purpose use or routine samples. Its soft HI polystyrene composition prevents the head breaking off, which makes it ideal for clinical use.
Brand:  Technical Service Consultants™ TS/1-C
 View more versions of this product
Product Code. 12769865
  • 237.00 € / Pack of 1000
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Laboratory use only
  • Shelf Life: 3 years.
  • Sterilization Method: Gamma Irradiation to achieve a SAL of 10-6
  • 4-25°C. Store in dry and ventilated area; protect from light, heat and moisture in tightly closed original container
Specifications
Specifications
Single-use
Polystyrene
White
Laboratory use only
Packaged in peel pouch
10 μL
Sterile
Inoculating Loop
1000 Pk.
Laboratory use only