missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contec™ Sterile IPA
Provided sterile by gamma irradiated using a validated process at no less than 25 kGy. Contec™ Sterile IPA is an alcohol blend of 0.2 micron filtered, filled and bagged in a Grade C (ISO Class 7) cleanroom.
Brand: Contec™ SBT0570IW
177.99 EUR valid until 2025-02-28
Use promo code "24154" to get your promotional price.
This item is not returnable.
View return policy
Description
- Guaranteed endotoxin levels of <0.25EU/mL make it ideal for use in product contact areas
- GMP manufactured under Grade A laminar flow air flow in a Grade C cleanroom ensures the alcohol, container and packaging are free from contamination and particulates
- Trigger spray and bag in bottle protected system prevents bottle contents from becoming contaminated during use
Specifications
Isopropyl Alcohol | |
Not Applicable | |
Colorless | |
Clear | |
70% IPA with water for injection | |
Limited-use | |
Yes | |
0.1 gal. |
Liquid Spray | |
Sterile | |
Not Autoclavable | |
Fluid | |
Trigger Spray Bottle | |
Filtered | |
8 x 0.5 L | |
0.5 L |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- CONTEC STERILE 70% IPA WITH WFI
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P370+P378-In case of fire: Use dry chemical, carbondioxide to extinguish.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : PROPAN-2-OL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction