Learn More
SP Bel-Art™ Sterileware Jumbo Sampling Pipettes
Ideal for sampling from narrow- or wide-mouth containers up to 208-liter (55 Gallon) drums
Brand:  SP Bel-Art™ F37872-1100
 View more versions of this product
Product Code. 16896916
  • 452.30 € / Pack of 37
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Reaches into container while keeping hand clear of contents
  • Diameter allows for use through drum bunghole or other small openings
  • Color-coded end helps prevent handling errors
  • Low-viscosity liquids enter easily through the 3 mm (1⁄8 in.) tip hole
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) pipettes heat sealed in individual polyethylene bags
  • Gamma irradiated sterile (SAL 10^-6)
  • Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Specifications
Specifications
225 mL
37/Pk.
2.1 cm
110 cm