Learn More
Product Code | Brand | Material | Dimensions (L x W) | No. per Case | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Dimensions (L x W) | No. per Case | Price | Quantity & Availability | ||||
11399093
![]() |
Thermo Scientific™
509-17 |
HDPE | 660 x 305mm | 200 |
95.95 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
||||
11309103
![]() |
Thermo Scientific™
|
HDPE | 610 x 406mm | 200 |
N/A
|
N/A
![]() |
||||
11359103
![]() |
Thermo Scientific™
510-17 |
HDPE | 660 x 305mm | 500 |
278.00 €
Pack of 500 |
Estimated Shipment: 24-06-2025
Log in to see stock. |
||||
11844145
![]() |
Thermo Scientific™
|
HDPE | 610 x 406mm | 500 |
N/A
|
N/A | ||||
11339103
![]() |
Thermo Scientific™
511-17 |
HDPE | 810 x 610mm | 200 |
292.00 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
||||
11379103
![]() |
Thermo Scientific™
509HT |
Polypropylene | 660 x 305mm | 200 |
192.00 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
||||
11349103
![]() |
Thermo Scientific™
510HT |
Polypropylene | 660 x 305mm | 500 |
452.00 €
Pack of 500 |
In Stock
Log in to see stock. |
||||
11389103
![]() |
Thermo Scientific™
509LHT |
Polypropylene | 610 x 406mm | 200 |
211.00 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
||||
11369103
![]() |
Thermo Scientific™
510LHT |
Polypropylene | 601 x 406mm | 500 |
516.00 €
Pack of 500 |
Estimated Shipment: 24-06-2025
Log in to see stock. |
||||
11329103
![]() |
Thermo Scientific™
|
Polypropylene | 810 x 610mm | 200 |
N/A
|
N/A
![]() |
||||
Description
Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.
- HDPE bags are suitable for sterilization at 121°C and polypropylene bags are suitable for the inactivation of particularly resistant biological waste at 135°C
- Convenient “tissue box” cartons dispense bags individually
- Coated wire stands or cardboard holders available for use with autoclave bags
Ordering Alerts:
Not available in all geographic regions
Your input is important to us. Please complete this form to provide feedback related to the content on this product.