missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Sterisart™ Universal, Gen 4

The Sterisart™ Universal Gen 4 transforms sterility testing workflows with remote data sharing and paperless documentation. Advanced reporting software simplifies workflow management and ensures compliance with strict regulatory standards, such as 21 CFR Part 11, through secure electronic signatures and audit trails. This maintains data integrity and supports audit readiness.

22705.00 € - 35365.00 €

Specifications

Display Type Touchscreen
Enclosure Rating IP65
Maximum Pressure 3.5 bar
Motor Speed 0 to 100 rpm
Reversible No
View More Specs

Products 3
Product Code Brand Model Includes Certifications/Compliance Price Quantity & Availability  
Product Code Brand Model Includes Certifications/Compliance Price Quantity & Availability  
18415640
View Documents
Sartorius™
16421-B
Basic Without Integrated Barcode Scanner No 21 CFR Part 11 Conformity
22705.00 €
1 pack
Please to purchase this item. Need a web account? Register with us today!
18425640
View Documents
Sartorius™
16421-E
Essential Integrated Barcode Scanner No 21 CFR Part 11 Conformity
26175.00 €
1 pack
Please to purchase this item. Need a web account? Register with us today!
18405640
View Documents
Sartorius™
16421-A
Advanced Integrated Barcode Scanner 21 CFR Part 11 Conformity
35365.00 €
1 pack
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Universal sterility testing pump designed for benchtop use or for integration in isolators.
  • Low profile and compact footprint.
  • Flexible, modular and open design
  • 316 L mirror finish stainless steel.
  • Sleek design for impeccable hygiene: easy disassembly and smooth contours.
  • Real-Time pressure detection in the sterility testing canisters.
  • 7 inch high performance touch screen display with integrated barcode scanner.
  • Built for 21 CFR Part 11 and EU GMP Annex 11: Advanced software for sterility testing featuring user access control, audit trail, secure data handling, electronic signatures, and reports.
Specifications

Specifications

Touchscreen
3.5 bar
No
IP65
0 to 100 rpm
100/240 V
Videos
SDS
Documents

Documents

Product Certifications