Learn More
Sampling Systems™ SteriWare™ Disposable Ladle
BIOPHARMA
Use straight from the bag without the need for costly cleaning. Sampling Systems™ SteriWare™ Disposable Ladle allows for efficient surface sampling of liquids.
Brand:  Sampling Systems™ 8053H-50S
 View more versions of this product
Product Code. 15889441
  • 76.80 € / Pack of 40
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • The one piece moulded design has been specially constructed to go through a standard lid bung
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No cross contamination
  • No cleaning required
Specifications
Specifications
HDPE
Single-use
White
50 mL
Sterile
48 mm