Learn More
Sampling Systems™ SteriWare™ Disposable Powder Spatula
BIOPHARMA
Easily sample and dispense quantities of powders. Sampling Systems™ SteriWare™ Disposable Powder Spatula is tough and comfortable to use with its specially designed rounded handle and robust blade.
91.05 € - 198.00 €
Specifications
Product Type Disposable Spatula
Includes 100 Spatulas
Width (Metric) 19 mm
View More Specs
Products 12
Product Code Brand Color Length (Metric) Material Sterility Price Quantity & Availability  
Product Code Brand Color Length (Metric) Material Sterility Price Quantity & Availability  
10762674
BIOPHARMA
View Documents
Sampling Systems™
8095A-150
White 230 mm Polystyrene Non-sterile
91.05 €
Pack of 100
Estimated Shipment: 24-12-2024
to see stock.
Add to basket
12792477
BIOPHARMA
View Documents
Sampling Systems™
8095A-250S
White 330 mm Polystyrene Sterile
176.00 €
Pack of 100
Estimated Shipment: 13-12-2024
to see stock.
Add to basket
12957945
BIOPHARMA
View Documents
Sampling Systems™
8095H-150
White 230 mm HDPE Non-sterile
105.00 €
Pack of 100
Estimated Shipment: 24-12-2024
to see stock.
Add to basket
15806171
BIOPHARMA
View Documents
Sampling Systems™
8095R-150S
Red 230 mm Polystyrene Sterile
180.00 €
Pack of 100
Estimated Shipment: 30-12-2024
to see stock.
Add to basket
15809471
BIOPHARMA
View Documents
Sampling Systems™
8095B-150S
Blue 230 mm Polypropylene Sterile
169.00 €
Pack of 100
Estimated Shipment: 17-12-2024
to see stock.
Add to basket
15816171
BIOPHARMA
View Documents
Sampling Systems™
8095R-250S
Red 330 mm Polystyrene Sterile
194.00 €
Pack of 100
Estimated Shipment: 13-12-2024
to see stock.
Add to basket
15819471
BIOPHARMA
View Documents
Sampling Systems™
Blue 330 mm Polypropylene Non-sterile
178.00 €
Pack of 100
Estimated Shipment: 10-01-2025
to see stock.
Add to basket
15829471
BIOPHARMA
View Documents
Sampling Systems™
8095B-250S
Blue 330 mm Polypropylene Sterile
198.00 €
Pack of 100
Estimated Shipment: 13-12-2024
to see stock.
Add to basket
15839471
BIOPHARMA
View Documents
Sampling Systems™
8095R-150
Red 230 mm Polystyrene Non-sterile
162.00 €
Pack of 100
Estimated Shipment: 13-12-2024
to see stock.
Add to basket
15849471
BIOPHARMA
View Documents
Sampling Systems™
Red 330 mm Polystyrene Non-sterile
175.00 €
Pack of 100
Estimated Shipment: 10-01-2025
to see stock.
Add to basket
15889461
BIOPHARMA
View Documents
Sampling Systems™
8095A-250
White 330 mm Polystyrene Non-sterile
143.00 €
Pack of 100
Estimated Shipment: 13-12-2024
to see stock.
Add to basket
15899461
BIOPHARMA
View Documents
Sampling Systems™
8095B-150
Blue 230 mm Polypropylene Non-sterile
99.10 €
Pack of 100
Estimated Shipment: 10-01-2025
to see stock.
Add to basket
Description
Description
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Suitable for use in ATEX Zones 0, 1 and 2
  • Available in 2 different sizes and a variety of colors
  • No cross contamination
  • No cleaning required
Specifications
Specifications
Disposable Spatula
100 Spatulas
19 mm