Learn More
Sampling Systems™ SteriWare™ Disposable Sample Spoons
BIOPHARMA
Sampling Systems™ SteriWare™ Disposable Sampling Spoons are a range of low cost, single use spoons that allow for quick and efficient surface sampling of powders and granules.
Brand:  Sampling Systems™ 8088A-03S
 View more versions of this product
Product Code. 12782467
  • 77.25 € / Pack of 100
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No cross contamination
  • No cleaning required
Specifications
Specifications
Spoon
3 mL
Sterile
173 mm
Polystyrene
Single-use
White