Learn More
Sampling Systems™ SteriWare™ Disposable Sample Spoons
BIOPHARMA
Sampling Systems™ SteriWare™ Disposable Sampling Spoons are a range of low cost, single use spoons that allow for quick and efficient surface sampling of powders and granules.
55.10 € - 144.00 €
Specifications
Product Type Spoon
Disposable Single-use
View More Specs
Products 7
Product Code Brand Capacity (Metric) Color Length (Metric) Sterility Price Quantity & Availability  
Product Code Brand Capacity (Metric) Color Length (Metric) Sterility Price Quantity & Availability  
12782467
BIOPHARMA
View Documents
Sampling Systems™
8088A-03S
3 mL White 173 mm Sterile
77.25 €
Pack of 100
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
15589634
BIOPHARMA
View Documents
Sampling Systems™
8088R-10S
10 mL Red 185 mm Sterile
144.00 €
Pack of 100
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
15849461
BIOPHARMA
View Documents
Sampling Systems™
8088A-03
3 mL White 173 mm Non-sterile
55.10 €
Pack of 100
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
15859461
BIOPHARMA
View Documents
Sampling Systems™
8088A-10
10 mL White 185 mm Non-sterile
55.10 €
Pack of 100
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
15869461
BIOPHARMA
View Documents
Sampling Systems™
8088B-10
10 mL Blue 185 mm Non-sterile
60.40 €
Pack of 100
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15879461
BIOPHARMA
View Documents
Sampling Systems™
8088B-10S
10 mL Blue 185 mm Sterile
78.25 €
Pack of 100
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
12544366
BIOPHARMA
View Documents
Sampling Systems™
8088A-10S
10 mL White 185 mm Sterile
80.85 €
Pack of 100
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
Description
Description
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No cross contamination
  • No cleaning required
Specifications
Specifications
Spoon
Single-use