missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ SteriWare™ Disposable Sample Spoons
BIOPHARMA

Sampling Systems™ SteriWare™ Disposable Sampling Spoons are a range of low cost, single use spoons that allow for quick and efficient surface sampling of powders and granules.

Brand:  Sampling Systems™ 8088R-10S

 View more versions of this product

Product Code. 15589634

  • 144.00 € / Pack of 100
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No cross contamination
  • No cleaning required
Specifications

Specifications

Spoon
10 mL
Sterile
100/Box
Polystyrene
Single-use
Red
185 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.