Learn More
Sampling Systems™ SteriWare™ Disposable Tweezers
Pick up both small and large items with a special dual action design. Sampling Systems™ SteriWare™ Disposable Tweezers are ideal for picking up items up to 25mm wide.
Brand:  Sampling Systems™ 8025H-115
 View more versions of this product
Product Code. 15839421
  • 65.00 € / Pack of 100
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No Cross Contamination
  • No Cleaning required
Specifications
Specifications
HDPE
115 mm
Non-Sterile Forceps
Non-sterile