missing translation for 'onlineSavingsMsg'
Learn More

Heidolph Instruments Stirring Bar Sets

Stirring bars available in oval and cylindrical shape sets

Brand:  Heidolph Instruments 509-55000-00

 View more versions of this product

Product Code. 10393073

  • 20.50 € / Pack of 3
Estimated Shipment: 10-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Oval Stir Bars:

  • Shape is similar to that of the flask and ensures complete mixing
  • Equipped with an oval magnetic core, more force is available than with a cylindrical magnetic core
  • For applications in:
    • Round bottom flasks e.g. for reaction mixtures

Cylindrical Stir Bars:

  • Smooth operating conditions at optimum contact area
  • Long lifetime
  • For applications in:
    • Beakers
    • Crystallizing dishes
    • Heating bathes
    • Other flat bottom vessels
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
30 x 10 mm
10 mm
PTFE
Oval
White
PTFE
30 mm
3/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Heidolph Instruments Stirring Bar Sets > Size: 30 x 10 mm; Oval

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.