Learn More
Fisherbrand™ Stainless Steel Stirring Shafts with Propellers
Excellent at producing axial flows in vessels from top to bottom. Fisherbrand™ Stainless Steel Stirring Shafts with Propellers are for use with overhead stirrer units.
Brand:  Fisherbrand™ 37020-9053
 View more versions of this product
Product Code. 15103007
Description
Description
  • Available in a range of sizes
  • Resistant to corrosion
Specifications
Specifications
Propellor, 63.5mm
1 Each
305mm
Stirring Shaft
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Stainless Steel Stirring Shafts with Propellers > Size: 305mm; Propellor: 63.5mm