Learn More
Fisherbrand™ Stainless Steel Stirring Shafts with Propellers
Excellent at producing axial flows in vessels from top to bottom. Fisherbrand™ Stainless Steel Stirring Shafts with Propellers are for use with overhead stirrer units.
Brand:  Fisherbrand™ 37020-9054
 View more versions of this product
Product Code. 15113007
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Available in a range of sizes
  • Resistant to corrosion
Specifications
Specifications
Propellor, 63.5mm
1 Each
457mm
Stirring Shaft