Learn More
Thermo Scientific™ Storage Canes
Store various cryostorage tubes in these storage canes, available in standard or stepped format to accommodate a variety of vial sizes and designs.
Brand: Thermo Scientific™ DS5020-0000
40.62 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Store Thermo Scientific™ Nalgene™ or Thermo Scientifc™ Nunc™ cryostorage tubes in Thermo Scientific™ Storage Canes, available in standard or stepped format to accommodate a variety of vial sizes and designs.
- Able to store tubes protected with Thermo Scientific™ CryoFlex™ tubing
- Compatible with color coders that attach to the end of the cane and can be written on for quick identification and inventory management
Specifications
Coder | |
Aluminum | |
100/Pk., 100/Cs. |
White | |
Cryo Cane Coder | |
Not Applicable |