Learn More
Thermo Scientific™ Store Image and Database Management Software, enterprise edition
Used for managing the images and data automatically transferred from a Thermo Scientific™ High Content Product
Brand:  Thermo Scientific™ S0100015
Product Code. 17139625
  • 67500.00 € / Each
Explore more special offers
Description
Description
  • Thermo Scientific™ Store Image and Database Management Software, Enterprise Edition, is used for managing the images and data automatically transferred from a Thermo Scientific™ High Content Product.
  • It allows for rapid, highly scalable, secure, storage of images and data with the ability to query.
  • Features of the Store Image and Database Management Software, Enterprise Edition:
  • Automatic Transfer of Images and Data—Centralized data storage allowing data and image access from multiple locations
  • Accessible and Linked —Data is seamlessly linked to its associated image
  • Visualize—View images and data linked together and easily compare images
  • export—Built in export formats for software such as Spotfire™, Excel™, De Novo™ Software, and GeneData Screener™ programs
  • The Store Image and Database Management Software maintains data and images on a SQL™ or Oracle™ database.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Image Management, Database Management
HCA/HCS
CellInsight
Data/Image Management
Enterprise
1 each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Store Image and Database Management Software, enterprise edition > 1 Ea.