Learn More
IBA Lifesciences Strep-Tactin™ Superflow™ High Capacity Column
Gravity flow column to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
Brand:  IBA Lifesciences 2-1209-550
 View more versions of this product
Product Code. 12856466
  • 329.00 € / Pack of 5
Estimated Shipment: 01-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Strep-Tactin™ Superflow™ high capacity columns contain a 6% agarose coupled with the streptavidin variant Strep-Tactin™ and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins via gravity flow.
  • In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™ Superflow™ high capacity has a higher density of immobilized Strep-Tactin™, which leads to a 3- to 5-fold higher yield of purified protein.
  • To reduce the waiting period during sample and buffer application, the use of the application aid WET FRED is recommended.
Specifications
Specifications
150 to 500 nmoL
Superflow 6
One-Step Purification of Recombinant Strep-Tag Proteins
1 mL
5 x 0.2 mL
Gravity Flow Strep-Tactin Superflow Column