Learn More
IBA Lifesciences Strep-Tactin™ Superflow™ High Capacity Column
Gravity flow column to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
144.00 € - 389.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12856466
|
IBA Lifesciences
2-1209-550 |
5 x 0.2 mL |
329.00 €
Pack of 5 |
Estimated Shipment: 01-10-2024
Log in to see stock. |
|||||
12806466
|
IBA Lifesciences
2-1209-001 |
1 mL |
144.00 €
1mL |
Estimated Shipment: 03-10-2024
Log in to see stock. |
|||||
12826466
|
IBA Lifesciences
2-1209-051 |
5 mL |
389.00 €
1mL |
Estimated Shipment: 01-10-2024
Log in to see stock. |
|||||
Description
- Strep-Tactin™ Superflow™ high capacity columns contain a 6% agarose coupled with the streptavidin variant Strep-Tactin™ and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins via gravity flow.
- In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™ Superflow™ high capacity has a higher density of immobilized Strep-Tactin™, which leads to a 3- to 5-fold higher yield of purified protein.
- To reduce the waiting period during sample and buffer application, the use of the application aid WET FRED is recommended.
For Research Use Only.