Learn More
IBA Lifesciences Strep-Tactin™XT 4Flow™ High Capacity Resin
Strep-Tactin™XT 4Flow™ high capacity resin is a 50% suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
188.00 € - 22575.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16674724
|
IBA Lifesciences
2-5030-002 |
4 mL |
188.00 €
1 set |
In Stock
Log in to see stock. |
|||||
16684724
|
IBA Lifesciences
2-5030-010 |
20 mL |
667.00 €
1 set |
In Stock
Log in to see stock. |
|||||
16694724
|
IBA Lifesciences
2-5030-025 |
50 mL |
1540.00 €
1 set |
In Stock
Log in to see stock. |
|||||
17277810
|
IBA Lifesciences
2-5030-500 |
1000 mL |
22575.00 €
1000mL |
Estimated Shipment: 31-12-2024
Log in to see stock. |
|||||
Description
- Strep-Tactin™XT 4Flow™ high capacity is a 4% agarose coupled with the streptavidin variant Strep-Tactin™XT and applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
- Due to the broad pH compatibility, excellent pressure stability, outstanding target purity (> 95%), and highest binding affinity, the resin is recommend for all protein classes, especially for low abundant or challenging proteins.
- The low concentrated agarose enables the simple attachment of larger proteins to the stationary phase, making it particularly suitable for purification of high-molecular-weight targets.
- In addition, Strep-Tactin™XT 4Flow™ high capacity allows the application of denaturing agents such as urea.
- In comparison to Strep-Tactin™XT 4Flow™, Strep-Tactin™XT 4Flow™ high capacity has a higher density of immobilized Strep-Tactin™XT, which leads to a 2-fold higher yield of purified protein.
- The suspension can be used for packing of gravity flow columns since the resin is optimized for column affinity chromatography, rather than for batch purification.
- To pack a column with 1 ml resin bed volume, 2 ml 50% suspension are necessary.