Learn More
IBA Lifesciences StrepMAB-Immo
StrepMAB-Immo is a monoclonal antibody to detect Strep-tag™II and Twin-Strep-tag™ fusion proteins
Brand: IBA Lifesciences 2-1517-001
This item is not returnable.
View return policy
Description
StrepMAB-Immo is a murine, high-affinity monoclonal antibody, which is especially suited for stable, mild, and oriented capturing of Strep-tag™II and Twin-Strep-tag™ fusion proteins on solid phases (e.g. Biacore CM5 sensor chips) or for detection of those proteins in applications like ELISA, FACS, immunohistochemistry, and immunocytochemistry. The nearly irreversible binding is validated for fusion proteins carrying a C- or N-terminal Strep-tag™II providing an N-terminal extension by a SerAla linker (recombinant protein-SA-WSHPQFEK or SA-WSHPQFEK-recombinant protein). Proteins containing the Strep-tag™II with N-terminal sequences other than SerAla are bound with reduced affinity in the cases tested so far, the Twin-Strep-tag™ always contains the SerAla linker (WSHPQFEK-GGGS-GGGS-GGSA-WSHPQFEK). Please note: For detection of Strep-tag™II or Twin-Strep-tag™ fusion proteins in western blots, the use of StrepMAB-Classic or Strep-Tactin™ is recommended.Specifications
Lyophilized powder | |
ELISA, FACS, Immunofluorescence, SPR | |
Suspension in PBS |
Monoclonal Antibody | |
-20°C | |
100 μg |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 10
Manual Irreversible-Biacore-Immobilization 2-1517-001
File size: 837.2 kB
Product Content Correction
Product Title
IBA Lifesciences StrepMAB-Immo > Volume: 100uL